Invitrogen
Manufacturer Code:PA554345
Catalog # PIPA554345
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human OBSCN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Obscurin; obscurin, myosin light chain kinase; Obscurin-MLCK; Obscurin-myosin light chain kinase; Obscurin-RhoGEF
Gene Aliases: ARHGEF30; KIAA1556; KIAA1639; OBSCN; UNC89
UniProt ID: (Human) Q5VST9
Entrez Gene ID: (Human) 84033
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.