Novus Biologicals
Manufacturer Code:NBP297423UV
Catalog # NB043275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human OBSCN fragment. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGEF30 DKFZp666E245 EC 2.7.11.1 FLJ14124 KIAA1556MGC120412 KIAA1639MGC138590 MGC120409 MGC120410 MGC120411 obscurin obscurin cytoskeletal calmodulin and titin-interacting RhoGEF obscurin myosin light chain kinase Obscurin-MLCK Obscurin-myosin light chain kinase obscurin-RhoGEF UNC89; Obscurin; obscurin, myosin light chain kinase; Obscurin-MLCK; Obscurin-myosin light chain kinase; Obscurin-RhoGEF
Gene Aliases: ARHGEF30; KIAA1556; KIAA1639; OBSCN; UNC89
UniProt ID: (Human) Q5VST9
Entrez Gene ID: (Human) 84033
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.