Novus Biologicals
Manufacturer Code:NBP198584
Catalog # NBP198584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is OBP2A - middle region. Peptide sequence NLEATFTFMREDRCIQKKILMRKTEEPGKFSAYGGRKLIYLQELPGTDDY. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOBPIIa OBP OBP2C OBPIIa odorant binding protein 2A odorant-binding protein 2a Odorant-binding protein IIa putative odorant-binding protein 2c; OBPIIa; Odorant-binding protein 2a; Odorant-binding protein IIa; putative odorant-binding protein 2c
Gene Aliases: hOBPIIa; LCN13; OBP; OBP2A; OBP2C; OBPIIa
UniProt ID: (Human) Q9NY56
Entrez Gene ID: (Human) 29991
Molecular Function: isomerase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.