Novus Biologicals
Manufacturer Code:NBP273140F
Catalog # NBP273140F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E11 |
Immunogen | Full length recombinant protein of human OBFC2B (NP_076973) produced in HEK293T cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: hSSB1; LP3587; LP3587 MGC2731 oligonucleotide/oligosaccharide-binding fold containing 2B Oligonucleotide/oligosaccharide-binding fold-containing protein 2B Sensor of single-strand DNA complex subunit B1 Sensor of ssDNA subunit B1 Single-stranded DNA-binding protein 1 SOSS complex subunit B1 SOSS-B1 SSB1hSSB1; Nucleic acid-binding protein 2; oligonucleotide/oligosaccharide-binding fold containing 2B; Oligonucleotide/oligosaccharide-binding fold-containing protein 2B; Sensor of single-strand DNA complex subunit B1; Sensor of ssDNA subunit B1; single strand DNA-binding protein 1; Single-stranded DNA-binding protein 1; SOSS complex subunit B1; SOSS-B1
Gene Aliases: hSSB1; LP3587; NABP2; OBFC2B; SOSS-B1; SSB1
UniProt ID: (Human) Q9BQ15
Entrez Gene ID: (Human) 79035
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.