Novus Biologicals
Manufacturer Code:NBP273138JF549
Catalog # N273138J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E4 |
Immunogen | Full length recombinant protein of human OBFC1 (NP_079204) produced in HEK293T cells. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AAF44 AAF-44 alpha accessory factor 44 bA541N10.2 CST complex subunit STN1 oligonucleotide/oligosaccharide-binding fold containing 1 Oligonucleotide/oligosaccharide-binding fold-containing protein 1 replication protein A 32 kDa subunit RP11-541N10.2 RPA-32 STN1FLJ22559 Suppressor of cdc thirteen homolog; alpha accessory factor 44; CST complex subunit STN1; oligonucleotide/oligosaccharide-binding fold containing 1; Oligonucleotide/oligosaccharide-binding fold-containing protein 1; replication protein A 32 kDa subunit; Suppressor of cdc thirteen homolog
Gene Aliases: AAF-44; AAF44; bA541N10.2; OBFC1; RPA-32; STN1
UniProt ID: (Human) Q9H668
Entrez Gene ID: (Human) 79991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.