Invitrogen
Manufacturer Code:PA575631
Catalog # PIPA575631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human OAZ3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antizyme 3; AZ3; OAZ-t; ODC-Az 3; Ornithine decarboxylase antizyme 3; Ornithine decarboxylase antizyme 3 AZ3 ODC-Az 3; testicular secretory protein Li 31; testis specific ornithine decarboxylase antizyme
Gene Aliases: Az-3; AZ3; OAZ-t; OAZ3; ODC-Az 3; TISP15
UniProt ID: (Human) Q9UMX2, (Mouse) Q9R109, (Rat) A1BPI0
Entrez Gene ID: (Human) 51686, (Mouse) 53814, (Rat) 689588
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.