Abnova
Manufacturer Code:H00051686K
Catalog # 89-201-787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human OAZ3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human OAZ3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: antizyme 3; AZ3; AZ3 OAZ-t TISP15; ODC-Az 3; Ornithine decarboxylase antizyme 3; testicular secretory protein Li 31
Gene Aliases: AZ3; OAZ-t; OAZ3; TISP15
UniProt ID: (Human) Q9UMX2
Entrez Gene ID: (Human) 51686
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.