Invitrogen
Manufacturer Code:PA514385
Catalog # PIPA514385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 2-31 amino acids from the N-terminal region of human OAZ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antizyme 1; AZ1; ODC-Az; Ornithine decarboxylase antizyme 1; Ornithine decarboxylase antizyme ODC-Az OAZ
Gene Aliases: Antizyme; AZ-1; AZ1; AZI; OAZ; OAZ1; ODC-Az
UniProt ID: (Human) O43382, (Mouse) P54369
Entrez Gene ID: (Human) 4946, (Mouse) 18245
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.