Invitrogen
Manufacturer Code:PA526615
Catalog # PIPA526615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 267-293 amino acids from the central region of human SLCO1B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HBLRR LST-1 LST1 OATP-C OATP1B1 OATP2 OATPC SLC21A6 OATP-2 liver-specific organic anion transporter 1 sodium-independent organic anion-transporting polypeptide 2 solute carrier family 21 (organic anion transporter) member 6 solute carrier family 21 member 6 solute carrier organic anion transporter family SLC01B1; Liver-specific organic anion transporter 1; LST-1; OATP-2; OATP-C; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; Solute carrier family 21 member 6; Solute carrier organic anion transporter family member 1B1; solute carrier organic anion transporter family, member 1B1
Gene Aliases: HBLRR; LST-1; LST1; OATP-C; OATP1B1; OATP2; OATPC; SLC21A6; SLCO1B1
UniProt ID: (Human) Q9Y6L6
Entrez Gene ID: (Human) 10599
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.