Invitrogen
Manufacturer Code:PA5106753
Catalog # PIPA5106753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLCO1B1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HBLRR Liver-specific organic anion transporter 1 LST1 LST-1 OATP1B1 OATP2 OATP-2 OATPC OATP-C SLC01B1 SLC21A6 SLCO1B1 sodium-independent organic anion-transporting polypeptide 2 solute carrier family 21 (organic anion transporter) member 6 Solute carrier family 21 member 6 Solute carrier organic anion transporter family member 1B1 solute carrier organic anion transporter family member 1B1; Liver-specific organic anion transporter 1; LST-1; OATP-2; OATP-C; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; Solute carrier family 21 member 6; Solute carrier organic anion transporter family member 1B1; solute carrier organic anion transporter family, member 1B1
Gene Aliases: HBLRR; LST-1; LST1; OATP-C; OATP1B1; OATP2; OATPC; SLC21A6; SLCO1B1
UniProt ID: (Human) Q9Y6L6
Entrez Gene ID: (Human) 10599
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.