Novus Biologicals
Manufacturer Code:NBP236492PE
Catalog # NBP236492PE
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E6.2F9 |
Immunogen | Two synthetic peptides made to the human OATP8 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5] |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Liver-specific organic anion transporter 2; Liver-specific organic anion transporter 2 liver-specific organic anion transporter 3TM13 LST2 LST-2 LST3 OATP1B3LST-3TM13 OATP8 OATP-8 Organic anion transporter 8 organic anion transporter LST-3c Organic anion-transporting polypeptide 8 SLC21A8 solute carrier family 21 (organic anion transporter) member 8 Solute carrier family 21 member 8 solute carrier organic anion transporter family member 1B3 solute carrier organic anion transporter family member 1B3; liver-specific organic anion transporter 3TM13; LST-2; OATP-8; Organic anion transporter 8; organic anion transporter LST-3c; Organic anion-transporting polypeptide 8; solute carrier family 21 (organic anion transporter), member 8; Solute carrier family 21 member 8; Solute carrier organic anion transporter family member 1B3; solute carrier organic anion transporter family, member 1B3
Gene Aliases: HBLRR; LST-2; LST-3TM13; LST2; LST3; OATP-8; OATP1B3; OATP8; SLC21A8; SLCO1B3
UniProt ID: (Human) Q9NPD5
Entrez Gene ID: (Human) 28234
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.