Invitrogen
Manufacturer Code:PA559067
Catalog # PIPA559067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human OAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GACR HOGA OATASE OKT gyrate atrophy ornithine aminotransferase mitochondrial ornithine delta-aminotransferase ornithine--oxo-acid aminotransferase ornithine-delta-aminotransferase ornithine-oxo-acid aminotransferase; gyrate atrophy; Ornithine aminotransferase, hepatic form; Ornithine aminotransferase, mitochondrial; Ornithine aminotransferase, renal form; Ornithine delta-aminotransferase; Ornithine--oxo-acid aminotransferase; ornithine-oxo-acid aminotransferase; testicular tissue protein Li 130
Gene Aliases: GACR; HOGA; OAT; OATASE; OKT
UniProt ID: (Human) P04181
Entrez Gene ID: (Human) 4942
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.