Invitrogen
Manufacturer Code:PA534873
Catalog # PIPA534873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 231 and 470 of OASL (Uniprot IDNo.Q15646) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2'-5'-OAS-related protein; 2'-5'-OAS-RP; 2'-5'-oligoadenylate synthase-like protein; 2'-5'-oligoadenylate synthetase-like; 59 kDa 2'-5'-oligoadenylate synthase-like protein; 59 kDa 2'-5'-oligoadenylate synthetase-like protein; OASLd TRIP-14 TRIP14 p59 OASL p59-OASL p59OASL 59 kDa 2&prime-5&prime-oligoadenylate synthase-like protein 59 kDa 2&prime-5&prime-oligoadenylate synthetase-like protein TR-interacting protein 14 thyroid receptor-interacting protein 14; p59 OASL; p59OASL; Thyroid receptor-interacting protein 14; TR-interacting protein 14
Gene Aliases: OASL; OASLd; p59 OASL; p59-OASL; p59OASL; TRIP-14; TRIP14
UniProt ID: (Human) Q15646
Entrez Gene ID: (Human) 8638
Molecular Function:
defense/immunity protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.