Novus Biologicals
Manufacturer Code:NBP294081100UL
Catalog # NB022721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 320-620 of human OAS3 (NP_006178.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (2-5&prime)oligo(A) synthase 32&prime-5&primeoligoadenylate synthetase p100 2-5A synthase 3 2&prime-5&prime-oligoadenylate synthase 3 2&prime-5&prime-oligoadenylate synthetase 3 100kDa P/OKcl.4 p100 OAS; (2-5')oligo(A) synthase 3; (2-5')oligo(A) synthetase 3; 2'-5'-oligoadenylate synthase 3; 2'-5'-oligoadenylate synthetase 3, 100kDa; 2'-5'oligoadenylate synthetase p100; 2-5A synthase 3; 2-5A synthetase 3; p100 OAS; p100OAS
Gene Aliases: OAS3; P/OKcl.4; p100; p100OAS
UniProt ID: (Human) Q9Y6K5
Entrez Gene ID: (Human) 4940
Molecular Function:
defense/immunity protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.