OriGene
Manufacturer Code:CF802825
Catalog # 50-166-6184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 60-335 of human OAS2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (2'-5')oligo(A) synthetase 2; (2-5')oligo(A) synthase 2; 2&prime-5&prime-oligoadenylate synthetase 2; 2'-5'-oligoadenylate synthase 2; 2'-5'-oligoadenylate synthetase 2, 69/71kDa; 2-5A synthase 2; p69 OAS / p71 OAS; p69OAS / p71OAS
Gene Aliases: OAS2
UniProt ID: (Human) P29728
Entrez Gene ID: (Human) 4939
Molecular Function:
defense/immunity protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.