Invitrogen
Manufacturer Code:PA543689
Catalog # PIPA543689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human OAS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (2-5')oligo(A) synthase 1; (2-5')oligo(A) synthase 1A; (2-5')oligo(A) synthetase 1; (2-5')oligo(A) synthetase 1A; 2&prime-5&prime-oligoadenylate synthetase 1 IFI-4 OIAS OIASI; 2',5'-oligo A synthetase 1; 2',5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5' oligoadenylate synthetase; 2'-5' oligoadenylate synthetase 1G; 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1A; 2'-5'-oligoadenylate synthetase 1; 2'-5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5'-oligoisoadenylate synthetase 1; 2-5-oligoadenylate synthetase 1; 2-5A synthase 1; 2-5A synthase 1A; 2-5A synthetase 1; 2-5A synthetase 1A; 25 oligoadenylate synthetase; E16 (2'-5') oligo A synthetase; E18/E16; p42 OAS; p46/p42 OAS
Gene Aliases: E18/E16; IFI-4; L3; OAS1; Oas1a; Oas1g; OIAS; Oias1; OIASI
UniProt ID: (Human) P00973, (Pig) Q29599, (Rat) Q05961, (Mouse) P11928
Entrez Gene ID: (Horse) 100034147, (Rabbit) 108178670, (Human) 4938, (Pig) 397570, (Dog) 608778, (Guinea pig) 100721851, (Rat) 192281, (Mouse) 246730
Molecular Function:
defense/immunity protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.