Invitrogen
Manufacturer Code:PA5115682
Catalog # PIPA5115682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human OAS1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (2-5')oligo(A) synthase 1; (2-5')oligo(A) synthase 1 (2-5')oligo(A) synthase 1A (2-5')oligo(A) synthetase 1 (2-5')oligo(A) synthetase 1A 2'5'-oligo A synthetase 1 2'5'-oligoadenylate synthetase 1 40/46kDa 25 oligoadenylate synthetase 2'-5' oligoadenylate synthetase 1A 2'-5' oligoadenylate synthetase 1G 2-5A synthase 1 2-5A synthase 1A 2-5A synthetase 1 2-5A synthetase 1A 2'-5'-oligoadenylate synthase 1 2'-5'-oligoadenylate synthase 1A 2'-5'-oligoadenylate synthetase 1 2'-5'-oligoadenylate synthetase 1 40/46kDa 2'-5'-oligoisoadenylate synthetase 1 E16 (2'-5') oligo A synthetase E18/E16 IFI 4 IFI4 IFI-4 L3 OAS 1 Oas1 Oas1a Oas1g OIAS Oias1 OIASI p42 OAS p46/p42 OAS; (2-5')oligo(A) synthase 1A; (2-5')oligo(A) synthetase 1; (2-5')oligo(A) synthetase 1A; 2',5'-oligo A synthetase 1; 2',5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5' oligoadenylate synthetase 1G; 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1A; 2'-5'-oligoadenylate synthetase 1; 2'-5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5'-oligoisoadenylate synthetase 1; 2-5A synthase 1; 2-5A synthase 1A; 2-5A synthetase 1; 2-5A synthetase 1A; 25 oligoadenylate synthetase; E16 (2'-5') oligo A synthetase; E18/E16; p42 OAS; p46/p42 OAS
Gene Aliases: E18/E16; IFI-4; L3; OAS1; Oas1a; Oas1g; OIAS; Oias1; OIASI
UniProt ID: (Human) P00973, (Rat) Q05961, (Mouse) P11928
Entrez Gene ID: (Human) 4938, (Rat) 192281, (Mouse) 246730
Molecular Function:
defense/immunity protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.