Novus Biologicals
Manufacturer Code:NBP294189100UL
Catalog # NB022935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 314-404 of human GPR143 (NP_000264.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 143 G-protein coupled receptor 143 NYS6 OA1Ocular albinism type 1 protein ocular albinism 1 (Nettleship-Falls); G-protein coupled receptor 143; ocular albinism 1; Ocular albinism type 1 protein
Gene Aliases: GPR143; NYS6; OA1
UniProt ID: (Human) P51810
Entrez Gene ID: (Human) 4935
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.