Invitrogen
Manufacturer Code:PA522830
Catalog # PIPA522830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide comprising residues 513-526 of the human Nurr1 protein. Based on 100&percnt sequence homology this antibody is also expected to cross react with rat and mouse Nurr1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HZF-3 NOT NURR1 RNR1 TINUR NGFI-B/nur77 beta-type transcription factor homolog T-cell nuclear receptor NOT immediate-early response protein NOT intermediate-early receptor protein nuclear receptor related 1 nuclear receptor subfamily 4 group A member 2 nur related protein-1 human homolog of orphan nuclear receptor NR4A2 orphan nuclear receptor NURR1 transcriptionally inducible nuclear receptor related 1 transcriptionally-inducible nuclear receptor; Immediate-early response protein NOT; intermediate-early receptor protein; NGFI-B/nur77 beta-type transcription factor homolog; Nuclear orphan receptor HZF-3; nuclear receptor related 1; Nuclear receptor subfamily 4 group A member 2; nuclear receptor subfamily 4 group A member 2 variant TINUR; nuclear receptor subfamily 4 group A member 2 variant NURR1a; nuclear receptor subfamily 4 group A member 2 variant NURR1b; nuclear receptor subfamily 4 group A member 2 variant NURR1c; nuclear receptor subfamily 4 group A member 2 variant NURR2c; nuclear receptor subfamily 4, group A, member 2; nur related protein 1; nur related protein-1, human homolog of; NUR-related factor 1; orphan nuclear receptor NR4A2; Orphan nuclear receptor NURR1; Regenerating liver nuclear receptor 1; RNR-1; SL-322; T-cell nuclear receptor NOT; transcriptionally inducible nuclear receptor related 1; Transcriptionally-inducible nuclear receptor
Gene Aliases: HZF-3; NOT; NR4A2; NURR1; RNR1; TINUR
UniProt ID: (Human) P43354, (Rat) Q07917
Entrez Gene ID: (Human) 4929, (Rat) 54278
Molecular Function: nuclear hormone receptor receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.