Novus Biologicals
Manufacturer Code:NBP299482F
Catalog # NB061119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Nurim. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NRM29Nuclear envelope membrane protein Nuclear rim protein nurim nurim (nuclear envelope membrane protein); Nuclear envelope membrane protein; Nuclear rim protein; Nurim
Gene Aliases: NRM; NRM29; UNQ555/PRO1112
UniProt ID: (Human) Q8IXM6
Entrez Gene ID: (Human) 11270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.