Invitrogen
Manufacturer Code:PA141391
Catalog # 01-675-520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 200-250 of human Nurim. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAAP-285E11.8 NRM29; Nuclear envelope membrane protein; Nuclear rim protein; Nurim
Gene Aliases: 2610307M02Rik; AI429796; NRM; NRM29; UNQ555/PRO1112
UniProt ID: (Human) Q8IXM6, (Mouse) Q8VC65
Entrez Gene ID: (Human) 11270, (Mouse) 106582
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.