Invitrogen
Manufacturer Code:PA537004
Catalog # PIPA537004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 399-444 of human NUP88 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; karyoporin; nuclear pore complex protein 88; Nuclear pore complex protein Nup88; Nuclear pore complex protein Nup88 88 kDa nucleoporin Nucleoporin Nup88; nucleoporin 88kDa; Nucleoporin Nup84; Nucleoporin Nup88; preimplantation protein 2
Gene Aliases: Nup84; NUP88; Prei2
UniProt ID: (Human) Q99567, (Rat) O08658, (Mouse) Q8CEC0
Entrez Gene ID: (Human) 4927, (Rat) 113929, (Mouse) 19069
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.