Novus Biologicals
Manufacturer Code:NB10093329
Catalog # NB10093329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human nucleoporin 153kDa using the numbering given in entry NP_005115.2 (GeneID 9972). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 153 kDa nucleoporin; CG4453 DmelCG4453 dmNup153 dNup153 HNUP153 N153 nuclear pore complex protein hnup153153 kDa nucleoporin nuclear pore complex protein Nup153 nucleoporin 153kD nucleoporin 153kDa Nucleoporin Nup153 Nup 153 nup153 Nup153 Nucleoporin 153; nuclear pore complex protein hnup153; Nuclear pore complex protein Nup153; nucleoporin 153kD; nucleoporin 153kDa; Nucleoporin Nup153
Gene Aliases: HNUP153; N153; NUP153
UniProt ID: (Human) P49790
Entrez Gene ID: (Human) 9972
Molecular Function: RNA binding protein nucleic acid binding structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.