Novus Biologicals
Manufacturer Code:NBP273104
Catalog # NBP273104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E11 |
Immunogen | Full length human recombinant protein of human NDEL1(NP_001020750) produced in HEk293 cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp451M0318 EOPA MITAP1mitosin-associated protein MITAP1 Mitosin-associated protein 1 nuclear distribution protein nudE-like 1 nudE nuclear distribution gene E homolog (A. nidulans)-like 1 nudE nuclear distribution gene E homolog like 1 nudE nuclear distribution gene E homolog like 1 (A. nidulans) NUDELendooligopeptidase A NUDE-like protein Protein Nudel; endooligopeptidase A; Mitosin-associated protein 1; mitosin-associated protein MITAP1; Nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog-like 1; Protein Nudel
Gene Aliases: EOPA; MITAP1; NDE1L1; NDE2; NDEL1; NUDEL
UniProt ID: (Human) Q9GZM8
Entrez Gene ID: (Human) 81565
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.