Invitrogen
Manufacturer Code:PA519082
Catalog # PIPA519082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CRNFAKDQASRKSY) corresponding to the internal amino acids of NDEL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: endooligopeptidase A; Mitosin-associated protein 1; mitosin-associated protein MITAP1; NDEL1 nudE nuclear distribution gene E homolog (A. nidulans)-like 1 DKFZp451M0318 EOPA MITAP1 NUDEL NUDE-like protein endooligopeptidase A mitosin-associated protein MITAP1 nudE nuclear distribution gene E homolog like 1; Nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog-like 1; Protein Nudel
Gene Aliases: EOPA; MITAP1; NDE1L1; NDE2; NDEL1; NUDEL
UniProt ID: (Human) Q9GZM8
Entrez Gene ID: (Human) 81565
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.