Invitrogen
Manufacturer Code:PA511440
Catalog # PIPA511440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 458-487 amino acids from the C-terminal region of human Nucleostemin (GNL3) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; guanine nucleotide binding protein-like 3 (nucleolar); Guanine nucleotide-binding protein-like 3; Guanine nucleotide-binding protein-like 3 Nucleolar GTP-binding protein 3 Nucleostemin E2-induced gene 3 protein Novel nucleolar protein 47 NNP47 E2IG3 NS; NNP47; Novel nucleolar protein 47; Nucleolar GTP-binding protein 3; Nucleostemin
Gene Aliases: C77032; E2IG3; GNL3; NNP47; NS
UniProt ID: (Human) Q9BVP2
Entrez Gene ID: (Human) 26354
Molecular Function:
G-protein
enzyme modulator
hydrolase
nucleic acid binding
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.