Proteintech
Manufacturer Code:180091AP150UL
Catalog # 50-173-1934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Nucleoside phosphorylase Fusion Protein Ag12507 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory sperm binding protein Li 156an; HEL-S-156an; Inosine phosphorylase; Inosine phosphorylase NP nucleoside phosphorylase PNP PRO1837 PUNP; inosine phosphorylase {ECO:0000250|UniProtKB:P00491}; Inosine-guanosine phosphorylase; nucleoside phosphorylase; PNP; PNP {ECO:0000250|UniProtKB:P00491}; Purine nucleoside phosphorylase; purine nucleoside phosphorylase {ECO:0000250|UniProtKB:P00491}; purine-nucleoside phosphorylase 1; purine-nucleoside:orthophosphate ribosyltransferase
Gene Aliases: AL024301; AU015798; NP; Np-1; Np-2; PNP; Pnp1; PRO1837; PUNP
UniProt ID: (Human) P00491, (Mouse) Q4FJT6, (Rat) P85973
Entrez Gene ID: (Human) 4860, (Mouse) 18950, (Rat) 290029
Molecular Function:
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.