Novus Biologicals
Manufacturer Code:NBP2446130.2MG
Catalog # NBP24461302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 364-5 + NCL/902 |
Immunogen | Lysate of SU-DHL-1 Nuclei (364-5) Recombinant human NCL protein (NCL/902) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C23 FLJ45706 FLJ59041 nucleolin Protein C23; Nucleolin; Protein C23
Gene Aliases: C23; NCL
UniProt ID: (Human) P19338
Entrez Gene ID: (Human) 4691
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.