Invitrogen
Manufacturer Code:MA535792
Catalog # PIMA535792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1504 |
Immunogen | A synthesized peptide derived from human Nuclear Matrix Protein p84 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 3110002N20Rik AW107452 BOS_22300 Hpr1 hTREX84 I79_014746 LRRGT00070 NMP-84 Nuclear matrix protein p84 P84 p84N5 THO complex subunit 1 Tho1 Thoc1; hTREX84; Nuclear matrix protein p84; p84N5; THO complex subunit 1; Tho1
Gene Aliases: 3110002N20Rik; AW107452; HPR1; NMP-84; P84; P84N5; THOC1
UniProt ID: (Human) Q96FV9, (Mouse) Q8R3N6
Entrez Gene ID: (Human) 9984, (Mouse) 225160
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.