Invitrogen
Manufacturer Code:PA316829
Catalog # APSA6829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full-length recombit human NuMA expressed in insect Sf9 cells using baculovirus expression system |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: centrophilin stabilizes mitotic spindle in mitotic cells; NMP-22; Nuclear matrix protein-22; Nuclear Mitotic Apparatus Protein; Nuclear mitotic apparatus protein 1; NuMA protein; SP-H antigen; structural nuclear protein
Gene Aliases: 6720401E04Rik; AA764025; AL022610; AU014979; NMP-22; NMP22; NUMA; NUMA1
UniProt ID: (Human) Q14980, (Mouse) E9Q7G0
Entrez Gene ID: (Human) 4926, (Rat) 308870, (Mouse) 101706
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.