Novus Biologicals
Manufacturer Code:NBP1778220.1MG
Catalog # NBP17782201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a purified recombinant mouse Nrf1 protein corresponding to aa 1- 534 of the native protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: Alpha palindromic-binding protein; Alpha palindromic-binding protein ALPHA-PAL EWG NRF-1 nuclear respiratory factor 1; Alpha-pal; NRF-1; Nuclear respiratory factor 1
Gene Aliases: ALPHA-PAL; NRF1
UniProt ID: (Human) Q16656
Entrez Gene ID: (Human) 4899
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.