Novus Biologicals
Manufacturer Code:NBP188365
Catalog # NBP188365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:REDYICYARFNHTQTIQQKQPISVKVISVDELNDTIAANLSDTEFYGAKSSRERPPTFLTPEGNASNKEELRGNVLSLECIAEGLPTPIIYWAKEDGMLPKNRTVYKNFEKTLQIIHVSEADSGNYQC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bravo hBravo KIAA0343Neuronal surface protein Bravo MGC138845 MGC138846 neuronal cell adhesion molecule Ng-CAM-related NgCAM-related cell adhesion molecule nr-CAM; hBravo; Neuronal cell adhesion molecule; Neuronal surface protein Bravo; Ng-CAM-related; NgCAM-related cell adhesion molecule; Nr-CAM
Gene Aliases: KIAA0343; NRCAM
UniProt ID: (Human) Q92823
Entrez Gene ID: (Human) 4897
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.