Abnova
Manufacturer Code:PAB17207
Catalog # 89-118-878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to N-terminus of mouse Nr2c2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Nr2c2.
For Research Use Only
Protein Aliases: Nuclear receptor subfamily 2 group C member 2; Orphan nuclear receptor TR4; orphan receptor TR4; Testicular receptor 4; Tr4; TR4 orphan receptor; TR4-NS orphan receptor
Gene Aliases: Nr2c2; Tr4
UniProt ID: (Rat) P55094
Entrez Gene ID: (Rat) 50659
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.