Abnova
Manufacturer Code:PAB7213
Catalog # 89-133-051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Npy2r. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Npy2r.
For Research Use Only
Protein Aliases: MGC124241; MGC124242; MGC124244; Neuropeptide Y receptor type 2; NPY-Y2 receptor; NPY2-R; Y2 receptor
Gene Aliases: NPY2-R; Npy2r
UniProt ID: (Mouse) P97295
Entrez Gene ID: (Mouse) 18167
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.