Novus Biologicals
Manufacturer Code:NBP259332C
Catalog # NBP259332C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S408-79 |
Immunogen | Fusion protein amino acids 597-802 (C-terminus) of rat Npas4. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: bHLHe79; BHLHE79 bHLHe79neuronal PAS domain-containing protein 4 Class E basic helix-loop-helix protein 79 Le-PAS neuronal PAS domain protein 4 Neuronal PAS4 NXFHLH-PAS transcription factor NXF PASD10PAS domain-containing protein 10; Class E basic helix-loop-helix protein 79; HLH-PAS transcription factor NXF; Neuronal PAS domain-containing protein 4; Neuronal PAS4; PAS domain-containing protein 10
Gene Aliases: BHLHE79; Le-PAS; NPAS4; NXF; PASD10
UniProt ID: (Human) Q8IUM7
Entrez Gene ID: (Human) 266743
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.