Novus Biologicals
Manufacturer Code:NBP225266SS
Catalog # NBP225266SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 69N15C9 |
Immunogen | This Noxa antibody (69N15C9) is made against amino acids 1-103 of mouse Noxa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: adult T cell leukemia-derived PMA-responsive; adult T cell leukemia-derived PMA-responsive APR Immediate-early-response protein APR NOXAPMA-induced protein 1 phorbol-12-myristate-13-acetate-induced protein 1 PMA-Induced Protein 1 Protein Noxa; Immediate-early-response protein APR; Phorbol-12-myristate-13-acetate-induced protein 1; PMA-induced protein 1; Protein Noxa
Gene Aliases: APR; NOXA; PMAIP1
UniProt ID: (Human) B2R4T7
Entrez Gene ID: (Human) 5366
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.