R&D Systems
Manufacturer Code:MAB8158100
Catalog # MAB8158100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 996715 |
Immunogen | <em>E. coli</em>-derived recombinant human NOX4 Lys450-Ser578 Accession No. Q9NPH5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Mouse Monoclonal Antibody
Protein Aliases: EC 1.6.3 Kidney oxidase-1 Kidney superoxide-producing NADPH oxidase KOX KOX-1 KOX-1Renal NAD(P)H-oxidase NADPH oxidase 4 RENOX RENOXEC 1.6.3.-; Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX-1; NADPH oxidase 4; Renal NAD(P)H-oxidase
Gene Aliases: KOX; KOX-1; NOX4; RENOX
UniProt ID: (Human) Q9NPH5
Entrez Gene ID: (Human) 50507
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.