Invitrogen
Manufacturer Code:12576380
Catalog # 50-108-50
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | HMN3-133 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: CADASIL CASIL; Neurogenic locus notch homolog protein 3; Notch 3; Notch 3 extracellular truncation; Notch 3 intracellular domain; Notch gene homolog 3
Gene Aliases: AW229011; hpbk; N3; Notch3
UniProt ID: (Mouse) Q61982
Entrez Gene ID: (Mouse) 18131
Molecular Function: defense/immunity protein membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.