Novus Biologicals
Manufacturer Code:NBP267420
Catalog # NBP267420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SJ205 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: EC 2.1.2.11 EC 3.4.21.68 hN1 Notch (Drosophila) homolog 1 (translocation-associated) notch 1Notch homolog 1 translocation-associated (Drosophila) Notch homolog 1 translocation-associated TAN1neurogenic locus notch homolog protein 1 Translocation-associated notch protein TAN-1; Neurogenic locus notch homolog protein 1; NEXT; NICD; Notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch homolog 1, translocation-associated; Translocation-associated notch protein TAN-1
Gene Aliases: AOS5; AOVD1; hN1; NOTCH1; TAN1
UniProt ID: (Human) P46531
Entrez Gene ID: (Human) 4851
Molecular Function:
defense/immunity protein
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.