Abnova
Manufacturer Code:MAB18456
Catalog # 89-338-381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S253-32 |
Immunogen | Recombinant protein corresponding to amino acids 20-216 at N-terminus of mouse Notch1. |
Conjugate | PCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 9930111A19Rik; lin-12; major type A protein; Mis6; Motch A; mT14; Neurogenic locus notch homolog protein 1; NEXT; NICD; Notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; p300; Tan1; transmembrane receptor Notch1
Gene Aliases: 9930111A19Rik; lin-12; Mis6; Motch; N1; Notch1; Tan1
UniProt ID: (Mouse) Q01705
Entrez Gene ID: (Mouse) 18128
Molecular Function:
defense/immunity protein
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.