OriGene
Manufacturer Code:CF500078
Catalog # 50-165-9347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3E12 |
Immunogen | Recombit protein expressed in E.coli corresponding to amino acids 2300-2556 of human NOTCH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AOS5 AOVD1 TAN1 hN1; Neurogenic locus notch homolog protein 1; NEXT; NICD; Notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch homolog 1, translocation-associated; Translocation-associated notch protein TAN-1
Gene Aliases: AOS5; AOVD1; hN1; NOTCH1; TAN1
UniProt ID: (Human) P46531
Entrez Gene ID: (Human) 4851
Molecular Function:
defense/immunity protein
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.