Novus Biologicals
Manufacturer Code:NBP266985
Catalog # NBP266985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM101-05 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Neurogenic locus notch homolog protein 4; neurogenic locus notch homolog protein 4 notch 4 Notch homolog 4 NOTCH3; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch homolog 4
Gene Aliases: INT3; NOTCH4
UniProt ID: (Human) Q99466
Entrez Gene ID: (Human) 4855
Molecular Function: defense/immunity protein membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.