R&D Systems
Manufacturer Code:AF3847
Catalog # AF3847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Notch-4 Intracellular Domain Gly1778-Lys2003 Accession # Q99466 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: INT3 neurogenic locus notch homolog protein 4 notch 4 Notch homolog 4 NOTCH3 Notch4; Neurogenic locus notch homolog protein 4; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch homolog 4
Gene Aliases: INT3; NOTCH4
UniProt ID: (Human) Q99466
Entrez Gene ID: (Human) 4855
Molecular Function: defense/immunity protein membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.