R&D Systems
Manufacturer Code:AF5267
Catalog # AF5267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Notch-1 Ala19-Gln526 Accession # AAM28905 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.2.11 EC 3.4.21.68 hN1 Notch (Drosophila) homolog 1 (translocation-associated) notch 1Notch homolog 1 translocation-associated (Drosophila) Notch homolog 1 translocation-associated Notch1 TAN1 TAN1neurogenic locus notch homolog protein 1 Translocation-associated notch protein TAN-1; Neurogenic locus notch homolog protein 1; NEXT; NICD; Notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch homolog 1, translocation-associated; Translocation-associated notch protein TAN-1
Gene Aliases: AOS5; AOVD1; hN1; NOTCH1; TAN1
UniProt ID: (Human) P46531
Entrez Gene ID: (Human) 4851
Molecular Function: defense/immunity protein membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.