Abnova
Manufacturer Code:MAB1470
Catalog # 89-110-448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NOS-B1 |
Immunogen | Recombinant protein corresponding to amino acids 1-181 of rat Nos1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100&mug/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant Nos1.
For Research Use Only
Protein Aliases: BNOS; Constitutive NOS; N-NOS; NC-NOS; Neuronal NOS; nitric oxidase synthase; Nitric oxide synthase, brain; nNOS; NOS type I; Peptidyl-cysteine S-nitrosylase NOS1
Gene Aliases: Bnos; Nos1
UniProt ID: (Rat) P29476
Entrez Gene ID: (Rat) 24598
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.