Abnova
Manufacturer Code:PAB26722
Catalog # 89-128-362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S852 of mouse Nos1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Nos1.
For Research Use Only
Protein Aliases: 2310005C01Rik; bNOS; Constitutive NOS; N-NOS; NC-NOS; Neuronal NOS; Nitric oxide synthase, brain; nNOS; NO; NOS type I; Nos-1; NOS-I; Peptidyl-cysteine S-nitrosylase NOS1
Gene Aliases: 2310005C01Rik; bNOS; N-NOS; NC-NOS; nNOS; NO; NOS; Nos-1; NOS-I; Nos1
UniProt ID: (Mouse) Q9Z0J4
Entrez Gene ID: (Mouse) 18125
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.