Novus Biologicals
Manufacturer Code:NBP177014
Catalog # NBP177014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human Norrin. The immunogen is located within the first 50 amino acids of Norrin. Amino Acid Squence: DTDSKTDSSFIMDSDPRR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EVR2 exudative vitreoretinopathy 2 (X-linked) FEVR ND Norrie disease (pseudoglioma) Norrie disease protein norrin X-linked exudative vitreoretinopathy 2 protein; Norrie disease (pseudoglioma); Norrie disease protein; Norrin; X-linked exudative vitreoretinopathy 2 protein
Gene Aliases: EVR2; FEVR; ND; NDP
UniProt ID: (Human) Q00604
Entrez Gene ID: (Human) 4693
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.