Invitrogen
Manufacturer Code:MA524647
Catalog # PIMA524647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL3063 |
Immunogen | Recombit protein corresponding to Human Norepinephrine Transporter |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NaCl-dependent norepinephrine transporter; NE transporter; NET; neurotransmitter transporter; neurotransmitter transporter, noradrenal; Norepinephrine transporter; Sodium-dependent noradrenaline transporter; Sodium-dependent noradrenaline transporter NET Solute carrier family 6 member 2 SLC6A2 NAT1 NET1 SLC6A5; solute carrier family 6 (neurotransmitter transporter), member 2; solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2; Solute carrier family 6 member 2; solute carrier family 6 member 5; transmembrane region 4..26 transmembrane region 55..77 transmembrane region 105..123 transmembrane region 136..157
Gene Aliases: NAT1; NE-T; NET; NET1; SLC6A2; SLC6A5
UniProt ID: (Human) P23975, (Mouse) O55192
Entrez Gene ID: (Human) 6530, (Rat) 83511, (Mouse) 20538
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.