Invitrogen
Manufacturer Code:PA542080
Catalog # PIPA542080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC6A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NaCl-dependent norepinephrine transporter; NE transporter; NET; neurotransmitter transporter; neurotransmitter transporter, noradrenal; Norepinephrine transporter; Sodium-dependent noradrenaline transporter; Solute carrier family 6 (neurotransmitter transporter noradrenalin) member 2 NAT1 NET NET1 SLC6A5; solute carrier family 6 (neurotransmitter transporter), member 2; solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2; Solute carrier family 6 member 2; solute carrier family 6 member 5; transmembrane region 4..26 transmembrane region 55..77 transmembrane region 105..123 transmembrane region 136..157
Gene Aliases: NAT1; NE-T; NET; NET1; NORADR; SLC6A2; SLC6A5
UniProt ID: (Bovine) P51143, (Human) P23975, (Mouse) O55192
Entrez Gene ID: (Bovine) 282363, (Horse) 100061346, (Rabbit) 100341516, (Human) 6530, (Dog) 487272, (Rat) 83511, (Mouse) 20538
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.