Invitrogen
Manufacturer Code:MA110028
Catalog # 01-670-285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 37C12 |
Immunogen | Yeast nuclear preparation. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NOP58 NOP6 YOR310C NOP58 Nucleolar protein 58 Nucleolar protein 5 Nucleolar protein 6; Nop6p; Nucleolar protein 5; Nucleolar protein 58; Nucleolar protein 6; RNA-processing protein NOP58
Gene Aliases: D1018; NOP5; NOP58; NOP6; O6108; YDL213C; YOR310C
UniProt ID: (Baker's yeast) Q12499, (Baker's yeast) Q07623
Entrez Gene ID: (Baker's yeast) 854487, (Baker's yeast) 851313
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.